Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

525480

Sigma-Aldrich

2,5-Dibromo-3-octylthiophene

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18Br2S
CAS Number:
Molecular Weight:
354.14
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

96%

refractive index

n20/D 1.548 (lit.)

bp

316 °C (lit.)

density

1.44 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCCCCCCCc1cc(Br)sc1Br

InChI

1S/C12H18Br2S/c1-2-3-4-5-6-7-8-10-9-11(13)15-12(10)14/h9H,2-8H2,1H3

InChI key

PJGBSUPPENVFAD-UHFFFAOYSA-N

Application

Conducting polymer precursor.

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Oligothiophenes are important organic electronic materials which can be produced using synthetic intermediates and Suzuki coupling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service