Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51920

Sigma-Aldrich

3-Heptene

technical, cis+trans, ≥95% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2CH=CHCH2CH3
CAS Number:
Molecular Weight:
98.19
Beilstein/REAXYS Number:
1719014
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

assay

≥95% (GC)

refractive index

n20/D 1.405

bp

95-96 °C (lit.)

density

0.702 g/mL at 20 °C (lit.)

SMILES string

CCC\C=C\CC

InChI

1S/C7H14/c1-3-5-7-6-4-2/h5,7H,3-4,6H2,1-2H3/b7-5+

InChI key

WZHKDGJSXCTSCK-FNORWQNLSA-N

pictograms

FlameHealth hazard

signalword

Danger

hcodes

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

19.4 °F - closed cup

flash_point_c

-7 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service