Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51592

Sigma-Aldrich

2-Heptadecanone

≥99.0% (GC)

Synonym(s):

Methyl pentadecyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO(CH2)14CH3
CAS Number:
Molecular Weight:
254.45
Beilstein/REAXYS Number:
1707134
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥99.0% (GC)

mp

47-50 °C

SMILES string

CCCCCCCCCCCCCCCC(C)=O

InChI

1S/C17H34O/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(2)18/h3-16H2,1-2H3

InChI key

TVTCXPXLRKTHAU-UHFFFAOYSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaime Martínez-Harms et al.
Chembiochem : a European journal of chemical biology (2018-04-27)
Despite increasing evidence for biosynthetic connections between flower pigments and volatile compounds, examples of such relationships in polymorphic plant species remains limited. Herein, color-scent associations in flowers from Papaver nudicaule (Papaveraceae) have been investigated. The spectral reflectance and scent composition
Examination of spray paints on plasters by the use of pyrolysis-gas chromatography/mass spectrometry for forensic purposes.
Milczarek JM and Zieba-Palus J.
Journal of Analytical and Applied Pyrolysis, 86(2), 252-259 (2009)
Essential oil of Filipendula hexapetala.
Pavlovic M, et al.
Chemistry of Natural Compounds, 43(2), 228-229 (2007)
Volatile flavor components of cooked kaorimai (scented rice, O. sativa japonica).
Yajima I, et al.
Agricultural and Biological Chemistry, 43(12), 2425-2429 (1979)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service