Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51432

Sigma-Aldrich

(S)-(−)-α-Ethylbenzylamine

≥95.0% (GC)

Synonym(s):

(S)-(−)-1-Phenylpropylamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13N
CAS Number:
Molecular Weight:
135.21
Beilstein/REAXYS Number:
2412016
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

assay

≥95.0% (GC)

optical activity

[α]20/D −20±2°, neat

optical purity

enantiomeric ratio: ≥99.0:1.0 (GC)

refractive index

n20/D 1.520

density

0.940 g/mL at 20 °C (lit.)

SMILES string

CC[C@H](N)c1ccccc1

InChI

1S/C9H13N/c1-2-9(10)8-6-4-3-5-7-8/h3-7,9H,2,10H2,1H3/t9-/m0/s1

InChI key

AQFLVLHRZFLDDV-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(−)-α-Ethylbenzylamine can be used:
  • To prepare optically active copolyacrylate polymers with sensing and chiroptical properties.[1]
  • As a chiral auxiliary in the preparation of enantioenriched indanone derived α-SCF2H β-keto esters.[2]

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

170.6 °F

flash_point_c

77 °C

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Asymmetric Electrophilic Difluoromethylthiolation of Indanone-Based β-Keto Esters Using Difluoromethanesulfonyl Hypervalent Iodonium Ylides
Gondo S, et al.
Molecules (Basel), 24(2), 221-221 (2019)
Synthesis and characterization of new optically active poly (acrylamide/methacrylurea-co-vinyl acetate) copolymers with dansyl units
Murariu M and Buruiana EC
Designed Monomers and Polymers, 18(2), 118-128 (2015)

Articles

Chiral amines have found widespread application in asymmetric synthesis serving, for instance, as chiral bases in enantioselective deprotonation reactions or being valuable substances for resolving racemic mixtures of acids.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service