Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

51204

Sigma-Aldrich

(−)-α-Gurjunene

≥97.0% (sum of enantiomers, GC)

Synonym(s):

(-)-alpha-Gurjunene, (2S,4R,7R,8R)-3,3,7,11-Tetramethyltricyclo[6.3.0.02.4]undec-1(11)-ene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24
CAS Number:
Molecular Weight:
204.35
Beilstein/REAXYS Number:
1939514
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]20/D −213±5°, neat

refractive index

n20/D 1.501

density

0.918 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@@H]2[C@H](C3=C(C)CCC13)C2(C)C

InChI

1S/C15H24/c1-9-6-8-12-14(15(12,3)4)13-10(2)5-7-11(9)13/h9,11-12,14H,5-8H2,1-4H3/t9-,11?,12-,14-/m1/s1

InChI key

SPCXZDDGSGTVAW-VXYFJCLZSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

230.0 °F - closed cup

flash_point_c

110 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service