Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

499900

Sigma-Aldrich

2-Formyl-3-thiopheneboronic acid

≥95%

Synonym(s):

2-Formyl-3-thienylboronic acid, 2-Formylthien-3-ylboronic acid, 2-Formylthiophen-3-ylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BO3S
CAS Number:
Molecular Weight:
155.97
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

assay

≥95%

mp

167-193 °C (lit.)

SMILES string

OB(O)c1ccsc1C=O

InChI

1S/C5H5BO3S/c7-3-5-4(6(8)9)1-2-10-5/h1-3,8-9H

InChI key

BBENFHSYKBYWJX-UHFFFAOYSA-N

Application

2-Formyl-3-thiopheneboronic acid can be used as:
  • A substrate in the palladium-Tedicyp catalyzed Suzuki coupling reaction with different aryl bromides.[1]
  • A starting material for the synthesis of 4H-thieno[2,3-c]-isoquinolin-5-one derivatives as PARP-1 inhibitors.[2]
  • A starting material for the preparation of phenanthro-dithiophene moieties having field-effect transistor properties.[3]

Reactant for:
  • Suzuki cross-coupling reactions
  • Preparation of boronic acid esters

Other Notes

Contains varying amounts of anhydride

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of biheteroaryl derivatives by tetraphosphine/palladium-catalysed Suzuki coupling of heteroaryl bromides with heteroarylboronic acids
Kondolff I, et al.
J. Mol. Catal. A: Chem., 269(1-2), 110-118 (2007)
Transistor Properties of 2, 7-Dialkyl-Substituted Phenanthro [2, 1-b: 7, 8-b′] dithiophene
Kubozono Y, et al.
Scientific reports, 6, 38535-38535 (2016)
Towards new neuroprotective agents: design and synthesis of 4H-thieno [2, 3-c] isoquinolin-5-one derivatives as potent PARP-1 inhibitors
Pellicciari R, et al.
Il Farmaco (Societa Chimica Italiana : 1989), 58(9), 851-858 (2003)

Articles

Suzuki-Miyaura cross-coupling reaction is extensively used in organic chemistry, polymer science, and pharmaceutical industries.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service