Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

477079

Sigma-Aldrich

2,3-Dihydro-1,4-dithiino[2,3-c]furan-5,7-dione

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4O3S2
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

mp

112-115 °C (lit.)

SMILES string

O=C1OC(=O)C2=C1SCCS2

InChI

1S/C6H4O3S2/c7-5-3-4(6(8)9-5)11-2-1-10-3/h1-2H2

InChI key

MXSSHXZXAAXCOW-UHFFFAOYSA-N

General description

2,3-Dihydro-1,4-dithiino[2,3-c]furan-5,7-dione is an anhydride of dicarboxylic acid.[1]

Application

2,3-Dihydro-1,4-dithiino[2,3-c]furan-5,7-dione (5,6-dihydro-1,4-dithiin-2,3-dicarboxylic anhydride, dhca, 3,6-Dithia-3,4,5,6-tetrahydrophthalic anhydride) may be used in the synthesis of the following:
  • photoluminescent three-dimensional (3-D) AgI coordination polymer[2]
  • 5,6-dihydro-1,4- dithiin-2,3-dicarboxylate[3]
  • CuII and MnII coordination complexes with 5,6-dihydro-1,4-dithiin-2,3-dicarboxylate as a primary building block[3]
  • 2-heteroaryl-4,7-dithia-4,5,6,7-tetrahydro-1,3-indandiones[1]
  • 3-arylmethylene-4,7-dithia-4,5,6,7-tetrahydrophthalides[4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phthalides and 1, 3-indandiones. LII. Reactions of 3, 6-dithia-3, 4, 5, 6-tetrahydrophthalic anhydride with methyl derivatives of nitrogen-containing heterocycles.
Krasnec L and Hrnciar P.
Chemical Papers, 28(1), 100-105 (1974)
A 3-D silver (I) polymer with 5, 6-dihydro-1, 4-dithiin-2, 3-dicarboxylate ligand, showing an unusual 6-connected (4 12. 6 3) 2 (4 9. 6 6) topology.
Hu M, et al.
Inorganic Chemistry Communications, 13(12), 1548-1551 (2010)
In situ syntheses, crystal structures and magnetic properties of CuII and MnII coordination assemblies based on a novel heteroalicyclic dicarboxylate tecton and N-donor co-ligands.
Liu CS, et al.
CrystEngComm, 14(1), 160-168 (2012)
Phthalides and 1,3-indandiones. XLIX. Preparation of 3-arylmethylene-4,7-dithia-4,5-6,7-tetrahydrophthalides and 2-aryl-4,7-dithia-4,5,6,7-tetrahydro-1,3-indandiones.
Hrnciar P.
Chemical Papers, 27(3) , 372-380 (1973)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service