Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

465801

Sigma-Aldrich

2,6-Di-tert-butyl-4-ethylphenol

99%

Synonym(s):

2,6-Bis(1,1-dimethylethyl)-4-ethylphenol, 2,6-Di-tert -butyl-p -ethylphenol, 4-Ethyl-2,6-di-tert -butylphenol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(C2H5)OH
CAS Number:
Molecular Weight:
234.38
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

99%

bp

272 °C (lit.)

mp

43-46 °C (lit.)

SMILES string

CCc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C16H26O/c1-8-11-9-12(15(2,3)4)14(17)13(10-11)16(5,6)7/h9-10,17H,8H2,1-7H3

InChI key

BVUXDWXKPROUDO-UHFFFAOYSA-N

Packaging

Packaged in glass bottles

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

221.0 °F - closed cup

flash_point_c

105.00 °C - closed cup

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service