Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

465240

Sigma-Aldrich

4-Bromo-2-fluorobenzonitrile

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(F)CN
CAS Number:
Molecular Weight:
200.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

69-72 °C (lit.)

SMILES string

Fc1cc(Br)ccc1C#N

InChI

1S/C7H3BrFN/c8-6-2-1-5(4-10)7(9)3-6/h1-3H

InChI key

HGXWRDPQFZKOLZ-UHFFFAOYSA-N

Application

4-Bromo-2-fluorobenzonitrile may be used in the preparation of heterocycles[1][2][3] and liquid crystals.[4][5]
4-Bromo-2-fluorobenzonitrile may be used in the preparation of:
  • 3-fluoro-4′-formylbiphenyl-4-carbonitrile[6]
  • (S)-6-{5-[2-amino-3-(1H-indol-3-yl)-propoxy]-pyridin-3-yl}-1H-indazol-3-ylamine[7]
  • 4-methoxycarbonyl-2-fluorobenzonitrile, via palladium-mediated methoxy carbonylation[8]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 37, 2885-2885 (1996)
Journal of Materials Chemistry, 5, 1125-1125 (1995)
Mol. Cryst. Liq. Cryst., 172, 165-165 (1989)
Silyl-directed, iridium-catalyzed ortho-borylation of arenes. A one-pot ortho-borylation of phenols, arylamines, and alkylarenes.
Boebel TA and Hartwig JF.
Journal of the American Chemical Society, 130(24), 7534-7535 (2008)
Salvatore D Lepore et al.
Organic letters, 5(1), 7-10 (2003-01-03)
In both solution and the solid phase, a variety of ketone oxime anions have been treated with 4-substituted-2-fluorobenzonitriles to give the corresponding nucleophilic aromatic substitution aryloxime adducts. Under aqueous acidic conditions, these adducts underwent cyclization to give the corresponding ketones.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service