Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

461415

Sigma-Aldrich

(S)-2-Phenyl-1-propanol

98%

Synonym(s):

(S)-β-Methylphenethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(C6H5)CH2OH
CAS Number:
Molecular Weight:
136.19
Beilstein/REAXYS Number:
2500176
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

assay

98%

optical activity

[α]22/D −17°, neat

refractive index

n20/D 1.526 (lit.)

bp

214 °C (lit.)

density

1 g/mL at 25 °C (lit.)

SMILES string

C[C@H](CO)c1ccccc1

InChI

1S/C9H12O/c1-8(7-10)9-5-3-2-4-6-9/h2-6,8,10H,7H2,1H3/t8-/m1/s1

InChI key

RNDNSYIPLPAXAZ-MRVPVSSYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

226.4 °F - closed cup

flash_point_c

108 °C - closed cup

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service