Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

460222

Sigma-Aldrich

Diisodecyl glutarate

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H21O2C(CH2)3CO2C10H21
CAS Number:
Molecular Weight:
412.65
EC Number:
MDL number:
UNSPSC Code:
12162002

grade

technical grade

refractive index

n20/D 1.449 (lit.)

bp

>260 °C (lit.)

solubility

hexane, toluene, ethanol and acetone: soluble
water and propylene glycol: insoluble

density

0.92 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCCCOC(=O)CCCC(=O)OCCCCCCCC(C)C

Application

Plasticizer

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service