Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

457426

Sigma-Aldrich

5-Iodo-2,4-dimethoxy­pyrimi­dine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7IN2O2
CAS Number:
Molecular Weight:
266.04
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

mp

71-75 °C (lit.)

λmax

277 nm

SMILES string

COc1ncc(I)c(OC)n1

InChI

1S/C6H7IN2O2/c1-10-5-4(7)3-8-6(9-5)11-2/h3H,1-2H3

InChI key

KNTMOGOYRYYUIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nucleic acid components and their analogues. XLIII. Synthesis of anomeric 5-iodo-2'-deoxyuridines.
Prystas M and Sorm F.
Collection of Czechoslovak Chemical Communications, 29(1), 121-130 (1964)
Synthesis of 5-(acylethynyl) uracils and their corresponding 2'-deoxyribonucleosides through palladium-catalysed reactions.
Kundu BG and Dasgupta SK.
Journal of the Chemical Society. Perkin Transactions 1, 21, 2657-2663 (1993)
Synthesis of nucleosides. 9. General synthesis of N-glycosides. I. Synthesis of pyrimidine nucleosides.
Niedballa U and Vorbruggen H.
The Journal of Organic Chemistry, 39(25), 3654-3660 (1974)
Palladium-catalysed carbomethoxyvinylation and thienylation of 5-iodo (bromo)-2, 4-dimethoxypyrimidine in water.
Basnak I, et al.
Tetrahedron Letters, 38(27), 4869-4872 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service