Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

456683

Sigma-Aldrich

Methyl 5-methoxysalicylate

98%

Synonym(s):

Methyl 2-hydroxy-5-methoxybenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(OH)CO2CH3
CAS Number:
Molecular Weight:
182.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

refractive index

n20/D 1.544 (lit.)

bp

235-240 °C (lit.)

density

1.223 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1cc(OC)ccc1O

InChI

1S/C9H10O4/c1-12-6-3-4-8(10)7(5-6)9(11)13-2/h3-5,10H,1-2H3

InChI key

DFNBGZODMHWKKK-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New class of 1, 3-benzoxazinones as potential central nervous system agents
Heindel ND and Schaeffer LA.
Journal of Medicinal Chemistry, 13(5), 981-983 (1970)
Photoinduced intramolecular proton transfer in methyl salicylates: a theoretical study with spectroscopic support.
Sanchez-Cabezudo M, et al.
Journal of Molecular Structure, 131(3), 277-277 (1985)
One-pot synthesis of xanthones and thioxanthones by the tandem coupling-cyclization of arynes and salicylates.
Zhao J and Larock RC.
Organic Letters, 7(19), 4273-4275 (2005)
Excited state proton transfer: A new feature in the fluorescence of methyl 5-chlorosalicylate and methyl 5-methoxysalicylate.
Acu?a AU, et al.
Journal of Photochemistry, 30(3), 339-352 (1985)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service