Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

455172

Sigma-Aldrich

(R)-(−)-1-Benzyl-3-hydroxypiperidine

97%

Synonym(s):

(R)-1-Benzyl-3-piperidinol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H17NO
CAS Number:
Molecular Weight:
191.27
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

optical activity

[α]24/D −12°, c = 1 in methanol

refractive index

n20/D 1.547 (lit.)

bp

275 °C (lit.)

density

1.07 g/mL at 25 °C (lit.)

SMILES string

O[C@@H]1CCCN(C1)Cc2ccccc2

InChI

1S/C12H17NO/c14-12-7-4-8-13(10-12)9-11-5-2-1-3-6-11/h1-3,5-6,12,14H,4,7-10H2/t12-/m1/s1

InChI key

UTTCOAGPVHRUFO-GFCCVEGCSA-N

pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service