Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

45341

Sigma-Aldrich

(−)-Epiglobulol

technical, ≥95% (sum of enantiomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein/REAXYS Number:
4176294
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22
Product 45341 is not currently sold in your country. Contact Technical Service

grade

technical

assay

≥95% (sum of enantiomers, GC)

optical activity

[α]20/D −38±3°, neat

refractive index

n20/D 1.496

density

0.97 g/mL at 20 °C (lit.)

Storage temp.

2-8°C

SMILES string

C[C@@H]1CC[C@@H]2[C@@H]1[C@H]3[C@@H](CC[C@]2(C)O)C3(C)C

InChI

1S/C15H26O/c1-9-5-6-10-12(9)13-11(14(13,2)3)7-8-15(10,4)16/h9-13,16H,5-8H2,1-4H3/t9-,10-,11-,12-,13-,15+/m1/s1

Inchi Key

AYXPYQRXGNDJFU-RWXDJMAFSA-N

General description

(−)-Epiglobulol is a naturally available, aromadendrane sesquiterpene.[1]

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Sesquiterpene occurring in the essential oil of many plants[2]

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gold (I) as an Artificial Cyclase: Short Stereodivergent Syntheses of (−)-Epiglobulol and (−)-4-beta, 7alpha-and (−)-4-alpha, 7alpha-Aromadendranediols.
Carreras J, et al.
Angewandte Chemie (International Edition in English), 53(19), 4896-4899 (2014)
F. Sorm
Prog. Chem. Org. Nat. Prod., 19, 1-1 (1961)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service