Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

446688

Sigma-Aldrich

Poly(indene-co-coumarone)

average Mn ~735, flakes, Generally contains less than 10% coumarone.

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

flakes

mol wt

average Mn ~735

iodine value

49

transition temp

softening point 130 °C (Ring and ball, ASTM E 28)

density

1.14 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Additives to adhesive, aluminum paints, varnishes, rotogravure inks and rubber compounds.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service