Recommended Products
assay
97%
form
solid
bp
178-181 °C/0.2 mmHg (lit.)
mp
50-56 °C (lit.)
functional group
hydroxyl
sulfone
SMILES string
Cc1ccc(cc1)S(=O)(=O)CCO
InChI
1S/C9H12O3S/c1-8-2-4-9(5-3-8)13(11,12)7-6-10/h2-5,10H,6-7H2,1H3
InChI key
QJFIXBNLKARINT-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Molecules (Basel, Switzerland), 14(12), 5247-5280 (2009-12-25)
New arylsulfonyl proton pump inhibitor (PPI) prodrug forms were synthesized. These prodrugs provided longer residence time of an effective PPI plasma concentration, resulting in better gastric acid inhibition.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service