Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

445754

Sigma-Aldrich

Poly(isobutyl methacrylate)

average Mw ~130,000

Synonym(s):

Isobutyl methacrylate polymer, Poly(isobutyl methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H14O2)n
CAS Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

mol wt

average Mw ~130,000

reduced viscosity

35-45 cm3/g(20 °C)(lit.)

acid number

≤1 mg KOH/g

transition temp

Tg 65 °C

solubility

alcohols and water: insoluble
aromatic and aliphatic hydrocarbons, esters, ketones, glycol ethers and chlorinated hydrocarbons: soluble

density

1.05 g/mL at 25 °C

SMILES string

CC(C)COC(=O)C(C)=C

InChI

1S/C8H14O2/c1-4-5-6-10-8(9)7(2)3/h2,4-6H2,1,3H3

InChI key

SOGAXMICEFXMKE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PiBMA can be prepared by free radical polymerization.[1]

Application

Binder in plastic coatings, aerosol paints, primers and printing inks
Glycidyl methacrylate grafted poly(isobutyl methacrylate) (PiBMA) microbeads can be used as super adsorbent beads for adsorption of heavy metal ions such as uranyl ions from aqueous solutions.[2] A study reports its use in the fabrication of polymer/polymer nanocomposites.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and properties of poly (isobutyl methacrylate?co?butanediol dimethacrylate?co?methacryl polyhedral oligomeric silsesquioxane) nanocomposites.
Li G Z, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 43(2), 355-372 (2005)
Janaina Habib Jorge et al.
Quintessence international (Berlin, Germany : 1985), 40(10), e93-100 (2010-02-18)
To evaluate the effect of microwave postpolymerization heat treatment and water storage time on the cytotoxicity of denture base and reline acrylic resins. Sample disks of 6 acrylic resins were fabricated under aseptic conditions (Kooliner, GC; Tokuyama Rebase II, Tokuyama
A G Zuccari et al.
Bio-medical materials and engineering, 7(5), 327-343 (1997-01-01)
Acrylic resins are commonly used in many dental applications; especially in the fabrication of provisional fixed partial dentures. Among noticed technical drawbacks associated with this material are unsatisfactory mechanical properties. Moreover, if acrylic resins are exposed to moist environment, water
D Tantbirojn et al.
European journal of oral sciences, 108(6), 564-568 (2001-01-12)
An amphiphilic coating is configured as a substantive film that has a tendency for an in-plane two-dimensional polymerization. This coating is hypothesized to protect enamel from in vitro acid decalcification, assessed through the following artificial caries model. Three regions on
The effects of two temporary crown materials on the dental pulp of monkeys (Macaca fascicularis).
G J Pearson et al.
International endodontic journal, 19(3), 121-124 (1986-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service