Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

442534

Sigma-Aldrich

Methylmercury(II) chloride

Cl ~13 %

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3HgCl
CAS Number:
Molecular Weight:
251.08
Beilstein/REAXYS Number:
3600218
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

powder

composition

Cl, ~13%

mp

170-173 °C (lit.)

SMILES string

C[Hg]Cl

InChI

1S/CH3.ClH.Hg/h1H3;1H;/q;;+1/p-1

InChI key

BABMCXWQNSQAOC-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Methylmercury(II) chloride [HgMe(Cl)] can be used in the synthesis methylmercury(II) complex of pyridine-2-carbaldehyde thiosemicarbazone , benzaldehyde-4,4-dimethylthiosemicarbazone , bis(alkynyl)oligothiophenes and -bithiazoles.

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and molecular structures of methyl and phenylmercury (II) complexes with benzaldehyde-4, 4-dimethylthiosemicarbazone
Lopez-Torres E and Mendiola MA
Journal of Organometallic Chemistry, 725(8), 28-33 (2013)
Weiran Zheng et al.
Evidence-based complementary and alternative medicine : eCAM, 2020, 8607931-8607931 (2020-07-01)
Guizhi Fuling capsule (GZFLc) is a modern preparation from traditional Chinese Medicine. Guizhi Fuling was first prescribed by Zhang Zhongjing almost two thousand years ago for the treatment of primary dysmenorrhea. It has also been used to treat uterine fibroids
Matthew D Rand et al.
Neurotoxicology, 81, 1-10 (2020-08-01)
Developmental methylmercury (MeHg) exposure can have lasting consequences on neural development and motor function across the lifespan. Recent evidence for MeHg targeting of myogenic pathways has drawn attention to the possibility that developing skeletal muscle plays a role in the
Dimethylthallium (III) and methylmercury (II) derivatives of pyridine-2-carbaldehyde thiosemicarbazone: synthesis and structure
Casas J, et al.
J. Chem. Soc., Dalton Trans., 1253-1259 (1993)
K Vojnits et al.
Current medicinal chemistry, 19(36), 6224-6232 (2012-12-19)
Traditional approaches in evaluating the hazard of drug candidates on the developing offspring are often time-consuming and cost-intensive. Moreover, variations in the toxicological response of different animal species to the tested substance cause severe problems when extrapolating safety dosages for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service