Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441198

Sigma-Aldrich

Boc-L-leucinol

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CH[NHCO2C(CH3)3]CH2OH
CAS Number:
Molecular Weight:
217.31
Beilstein/REAXYS Number:
4178460
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

95%

form

liquid

optical activity

[α]23/D −27°, c = 2 in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

CC(C)C[C@@H](CO)NC(=O)OC(C)(C)C

InChI

1S/C11H23NO3/c1-8(2)6-9(7-13)12-10(14)15-11(3,4)5/h8-9,13H,6-7H2,1-5H3,(H,12,14)/t9-/m0/s1

InChI key

LQTMEOSBXTVYRM-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Precursor to enantiomerically pure α-methyl amines.[1] Starting material for the synthesis of enantiopure homo-β-amino acids[2][3] and N-protected α-amino aldehydes.[4] Used in the total synthesis of the gastro-protective amicoumacin C.[5]

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soucek, M. Urban, J.
Collection of Czechoslovak Chemical Communications, 60, 693-693 (1995)
Donner, B.G.
Tetrahedron Letters, 36, 1223-1223 (1995)
R.A. Ward et al.
Tetrahedron, 51, 12301-12301 (1995)
Caputo, R. et al.
Tetrahedron, 51, 12337-12337 (1995)
R. Caputo et al.
Tetrahedron Letters, 36, 167-167 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service