Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

438421

Sigma-Aldrich

Silver cyanide

99.96% trace metals basis

Synonym(s):

Silver monocyanide 2, Silver(I) cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgCN
CAS Number:
Molecular Weight:
133.89
Beilstein/REAXYS Number:
4360924
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
assay:
99.96% trace metals basis
form:
powder

assay

99.96% trace metals basis

form

powder

impurities

≤500.0 ppm Trace Metal Analysis

SMILES string

[Ag]C#N

InChI

1S/CN.Ag/c1-2;

InChI key

MFIHOCAEOJNSOL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in preparation of Tl[Ag(CN)2], which exhibits previously unknown Ag(CN)2 luminescence.[1]

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Skin Irrit. 2

supp_hazards

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammad A. Omary et al.
Inorganic chemistry, 37(6), 1380-1386 (2001-10-24)
The structure of thallium dicyanoargentate(I) has been determined crystallographically. The crystal structure shows an Ag-Ag distance of 3.11 Å. This is the shortest Ag-Ag distance reported for any silver dicyanide salt whose crystal structure has been determined. Raman spectra of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service