Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

435511

Sigma-Aldrich

Ethylenediamine tetrakis(propoxylate-block-ethoxylate) tetrol

average Mn ~3,600

Synonym(s):

Tetronic 701

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2N[(-CH2CH(CH3)O-)x[-CH2CH2O-]yH]2]2
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

mol wt

average Mn ~3,600

refractive index

n20/D 1.458

surface tension

36 dyn/cm, 25 °C, 0.1 wt. % in H2O

viscosity

600 cP(25 °C, Brookfield)(lit.)

transition temp

softening point −21 °C
cloud point 18 °C (1 wt. % aqueous solution)

density

1.02 g/mL at 25 °C

HLB

1 - 7

InChI

1S/C34H72N2O12/c1-27(45-23-31(5)41-15-11-37)19-35(20-28(2)46-24-32(6)42-16-12-38)9-10-36(21-29(3)47-25-33(7)43-17-13-39)22-30(4)48-26-34(8)44-18-14-40/h27-34,37-40H,9-26H2,1-8H3

InChI key

BFKFABWTAFNFID-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terminal secondary tetrahydroxy-functional oligomer.

Application

Hard and soft surface cleaners, defoamers in coatings and water treatment. Lubricant in metal working, anti-foaming aid and extender for linear and cross-linked polyesters and polyurethanes.

Features and Benefits

Surface activity is dependent on the EO/PO ratio. Lower foaming, greater defoaming and reduced gelling tendencies relative to Pluronic® surfactants. Terminal secondary hydroxyl groups have lower reactivity and acidity than the primary hydroxyl groups in Pluronic® surfactants.

Legal Information

Pluronic is a registered trademark of BASF

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Sens. 1B

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

>482.0 °F - closed cup

flash_point_c

> 250 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service