Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434817

Sigma-Aldrich

Poly(vinyl phenyl ketone)

pellets

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH2CH(COC6H5)-]n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

pellets

acid number

<0.1 mg KOH/g

transition temp

Tg 58 °C

solubility

esters, ketones, glycol ethers and aromatics: soluble

density

1.19 g/mL at 25 °C (lit.)

SMILES string

C=CC(=O)c1ccccc1

Application

Component of clear and pigmented lacquers.

Features and Benefits

Neutral, unsaponifiable ketone polymer.
Promotes adhesion in coatings. Improves gloss of coatings. Improves pigment loading in cellulosenitrate coatings. Increases hardness of adhesives containing soft polymers.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service