Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434612

Sigma-Aldrich

Vinyl acrylate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH=CH2
CAS Number:
Molecular Weight:
98.10
EC Number:
MDL number:
UNSPSC Code:
12162002

assay

98%

contains

200-400 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.432 (lit.)

bp

90-92 °C (lit.)

density

0.942 g/mL at 25 °C (lit.)

SMILES string

C=COC(=O)C=C

InChI

1S/C5H6O2/c1-3-5(6)7-4-2/h3-4H,1-2H2

InChI key

BLCTWBJQROOONQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service