Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

430161

Sigma-Aldrich

Poly[4,4′-methylenebis(phenyl isocyanate)-alt-1,4-butanediol/polytetrahydrofuran]

pellets, MDI-polyether polyurethane.

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C15H10N2O2 · C4H10O2 · (C4H8O)nH2O)x
CAS Number:
MDL number:
UNSPSC Code:
12162002

abrasion resistance

20 mg (Taber, H-22 wheel)

form

pellets

melt index

40 g/10 min (224°C/1.2kg)

hardness

82 (Shore A, ASTM D 2240)

transition temp

Tg −40 °C (DSC)
softening point 85 °C (Vicat, ASTM D 1525)

density

1.13 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Automotive fascia, cable jackets, side trim, fabric laminates, health care tubing, catheters, transdermal patches and in seals casters, shoes, bushings and wheels.

Features and Benefits

Tough, high-performance, wear-resistant elastomer with good clarity, chemical resistance, flexibility and toughness. Can be used in current form or further polymerized.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service