Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

429813

Sigma-Aldrich

(−)-N-Boc-D-α-phenylglycinol

99%

Synonym(s):

(R)-(−)-2-(Boc-amino)-2-phenylethanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CNHCH(C6H5)CH2OH
CAS Number:
Molecular Weight:
237.29
Beilstein/REAXYS Number:
4688248
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

99%

form

solid

optical activity

[α]19/D −38°, c = 1 in chloroform

optical purity

ee: 99% (GLC)

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

137-139 °C (lit.)

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CO)c1ccccc1

InChI

1S/C13H19NO3/c1-13(2,3)17-12(16)14-11(9-15)10-7-5-4-6-8-10/h4-8,11,15H,9H2,1-3H3,(H,14,16)/t11-/m0/s1

InChI key

IBDIOGYTZBKRGI-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used for the synthesis of homochiral N-protected β-amino sulfoxides[1] and α-amino acids.[2] Chiral synthon, which undergoes heterocyclizations.[3] Precursor to chiral oxazolidinones.[4]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Siedlecka, R. Skarzewski, J.
Synlett, 757-757 (1996)
Agami, C. et al.
Tetrahedron Letters, 34, 4509-4509 (1993)
Agami, C. et al.
Bulletin de la Societe Chimique De France, 132, 808-808 (1995)
Cox, G.G. Harwood, L.M.
Tetrahedron Asymmetry, 5, 1669-1669 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service