Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

423475

Sigma-Aldrich

Polyethylenimine, 80% ethoxylated solution

35-40 wt. % in H2O, average Mw ~70,000

Synonym(s):

PEI

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

mol wt

average Mw ~70,000

concentration

35-40 wt. % in H2O

refractive index

n20/D 1.413

pH

12

transition temp

pour point 0 °C

density

1.08 g/mL at 25 °C

Looking for similar products? Visit Product Comparison Guide

Application

Additive to coatings and printing inks, brightener in electroplating, cationic modifier in clays and starches and adhesion/substantivity promoter.

Features and Benefits

Base polymer
Non-yellowing, film forming polyamine. Highly branched, symmetrical polymer. About 80% of the primary and secondary amines are ethoxylated.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service