Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

418803

Sigma-Aldrich

tert-Amyl-tert-butylamine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C(CH3)2NHC(CH3)3
CAS Number:
Molecular Weight:
143.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

96%

refractive index

n20/D 1.421 (lit.)

bp

144 °C (lit.)

density

0.77 g/mL at 25 °C (lit.)

SMILES string

CCC(C)(C)NC(C)(C)C

InChI

1S/C9H21N/c1-7-9(5,6)10-8(2,3)4/h10H,7H2,1-6H3

InChI key

KZOPSPQZLMCNPF-UHFFFAOYSA-N

General description

tert-Amyl-tert-butylamine is a sterically hindered secondary alkyl amine. Its electrosynthesis, via one-pot ecofriendly method has been proposed. Electrochemical behavior of tert-amyl-tert-butylamine has been investigated.[1]

pictograms

FlameCorrosion

signalword

Danger

hcodes

Hazard Classifications

Flam. Liq. 2 - Skin Corr. 1B

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

71.6 °F - closed cup

flash_point_c

22 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Iluminada Gallardo et al.
The Journal of organic chemistry, 73(17), 6647-6656 (2008-07-30)
The electrosynthesis of highly hindered secondary alkyl diamines through the formation of C-N bonds has been successfully performed in one-pot under mild conditions using an environmentally friendly method. The electrochemical behavior of four hindered secondary alkyl amines (di-tert-butylamine (1), tert-amyl-tert-butylamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service