Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

418048

Sigma-Aldrich

Poly(isophorone diisocyanate) solution

70 wt. % in propylene glycol methyl ether acetate/xylene (1:1)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

concentration

70 wt. % in propylene glycol methyl ether acetate/xylene (1:1)

refractive index

n20/D 1.5

bp

~140 °C

density

1.08 g/mL at 25 °C

SMILES string

CC1(C)CC(CC(C)(C1)N=C=O)N=C=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service