Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

416401

Sigma-Aldrich

Poly(maleic acid-co-olefin) sodium salt solution

average Mw ~12,000, 25 wt. % in H2O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(CO2R)CH(CO2R)-]x[-CH2CH(R')-]y, R=H or Na, R'=-CH2(CH2)nCH3
MDL number:
UNSPSC Code:
12162002

mol wt

average Mw ~12,000

concentration

25 wt. % in H2O

refractive index

n20/D 1.384

density

1.29 g/mL at 25 °C

SMILES string

[Na]OC(=O)\C=C\C(=O)O[Na]


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service