Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

3DXCNT002

Sigma-Aldrich

3DXNANO ESD CNT-ABS carbon nanotube reinforced ABS 3D printing filament

greener alternative

black, diam. 2.85 mm

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352103
NACRES:
NA.23

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

diam.

2.85 mm

weight

1 kgper each reel

color

black

greener alternative category

InChI

1S/C8H8.C4H6.C3H3N/c1-2-8-6-4-3-5-7-8;1-3-4-2;1-2-3-4/h2-7H,1H2;3-4H,1-2H2;2H,1H2

InChI key

XECAHXYUAAWDEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3DXNano ESD ABS Carbon Nanotube (CNT) enhanced compounds are used in critical applications that require electrostatic discharge (ESD) protection and a high level of cleanliness. Manufactured with consistency in mind - 3DXTech starts with MG-94 Premium ABS and then compounded according to a proprietary formulation of multi-wall carbon nanotubes and process / dispersion modifiers , giving you a filament with excellent printing characteristics and consistent ESD properties.

Benefits of CNTs include:

  • Consistent surface resistivity
  • low additive loading rates
  • improved retention of impact & elongation\
  • low particulate contamination
  • minimal cntribution to outgassing and ionic contamination
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product belongs to Enabling category of greener alternatives thus aligns with "Design for energy efficency". Carbon reinforced ABS 3D printing filament has high tensile strength and low thermal expansion. Click here for more information.

Application

Typical Applications Include:

  • Semi-con: HDD Components; Wafer Handling; Jigs;Casings; & Connectors
  • Industiral: Converying;Metering; and Sensing applications
This product can be used in material extrusion 3D printing technique.

Preparation Note

Recommended Print Conditions:

  • Extruder: 245- 255 °C (Desktop printers). 3DXNano ESD ABS is a filled product and has a higher melt viscosity vs. unfilled ABS. Therefore; it is sometimes necessary to print at higher temperatures to allow the resin to flow properly.
  • Note for Stratasys users: This product works very well in a wide variety of Stratasys printers, assuming you′ve ifgured out a work-around on the chip issue. The STratasys extruder setting of 300 °C is a n artifact of how and where they place the thermocouples. ABS doe snot process at 300 °C in the "real work".
  • Bed Temp: 100-110 °C
  • Bed Prep: Kapton Tape; ABS/Acetone Slurry.

Legal Information

3DXNano is a trademark of 3DXTech

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

3D printing is a type of additive manufacturing that can be used to rapidly fabricate components with highly customizable geometries.

Three-dimensional printing technology applications range from personal tools to aerospace equipment.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service