Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

399922

Sigma-Aldrich

Dequalinium chloride

90%

Synonym(s):

1,1′-Decamethylenebis(4-aminoquinaldinium) dichloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H40Cl2N4
CAS Number:
Molecular Weight:
527.57
EC Number:
MDL number:
UNSPSC Code:
12352203

assay

90%

mp

≥300 °C (lit.)

SMILES string

[Cl-].[Cl-].Cc1cc(N)c2ccccc2[n+]1CCCCCCCCCC[n+]3c(C)cc(N)c4ccccc34

InChI

1S/C30H38N4.2ClH/c1-23-21-27(31)25-15-9-11-17-29(25)33(23)19-13-7-5-3-4-6-8-14-20-34-24(2)22-28(32)26-16-10-12-18-30(26)34;;/h9-12,15-18,21-22,31-32H,3-8,13-14,19-20H2,1-2H3;2*1H

InChI key

LTNZEXKYNRNOGT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amaury Graulich et al.
Journal of medicinal chemistry, 50(21), 5070-5075 (2007-09-18)
Starting from the scaffold of N-methyllaudanosine and N-methylnoscapine, which are known small conductance Ca2+-activated K+ channel blockers, original bis-isoquinolinium derivatives were synthezised and evaluated using binding studies, electrophysiology, and molecular modeling. These quaternary compounds are powerful blockers, and the most
Clarissa K L Ng et al.
Bioorganic & medicinal chemistry, 15(10), 3422-3429 (2007-03-27)
A series of bis(pyridinium)alkanes have been prepared and their antifungal activity, haemolytic activity and ability to inhibit fungal phospholipase B1 have been investigated, together with those of the commercially available antiseptics octenidine and dequalinium. Removal of the amino substituents from
Ulrik S Sørensen et al.
Journal of medicinal chemistry, 51(23), 7625-7634 (2008-11-13)
Small conductance Ca2+-activated K+ channels (SK channels) participate in the control of neuronal excitability, in the shaping of action potential firing patterns, and in the regulation of synaptic transmission.SK channel inhibitors have the potential of becoming new drugs for treatment
Amaury Graulich et al.
Bioorganic & medicinal chemistry letters, 18(11), 3440-3445 (2008-04-26)
So far, small conductance Ca(2+)-activated K(+) channel (SK) blockers mostly consist of quaternary ammonium derivatives or peptides. Due to their physicochemical properties, these blockers are not suitable to study the physiological roles of SK channels in the central nervous system
David I Fletcher et al.
Bioorganic & medicinal chemistry, 15(16), 5457-5479 (2007-06-15)
The synthesis and pharmacological testing of a series of non-peptidic blockers of the SK(Ca) (SK-3) channel is described. Target compounds were designed to mimic the spatial relationships of selected key residues in the energy-minimised structure of the octadecapeptide apamin, which

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service