Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

398292

Sigma-Aldrich

Palladium(II) cyanide

99.9% trace metals basis

Synonym(s):

Dicyanopalladium, Palladium dicyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd(CN)2
CAS Number:
Molecular Weight:
158.45
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
assay:
99.9% trace metals basis
form:
powder

assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: palladium

impurities

≤2000 ppm Trace metallic impurities analysis (ICP)

SMILES string

N#C[Pd]C#N

InChI

1S/2CN.Pd/c2*1-2;

InChI key

WDXPAFYMKZQTHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Effective catalyst for the quantitative and stereospecific ring opening of oxiranes.[1]

pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

supp_hazards

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Imi, K. et al.
The Journal of Organic Chemistry, 52, 1013-1013 (1987)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service