391883
Triethyl 3-phosphonopropionate
98%
Synonym(s):
3-Phosphonopropionic acid triethyl ester, Diethyl (2-ethoxycarbonylethyl)phosphonate, Ethyl 3-(diethoxyphosphinyl)propionate, NSC 133823
Select a Size
About This Item
Recommended Products
assay
98%
form
liquid
reaction suitability
reaction type: C-C Bond Formation
refractive index
n20/D 1.433 (lit.)
bp
123-124 °C/3 mmHg (lit.)
density
1.094 g/mL at 25 °C (lit.)
SMILES string
CCOC(=O)CCP(=O)(OCC)OCC
InChI
1S/C9H19O5P/c1-4-12-9(10)7-8-15(11,13-5-2)14-6-3/h4-8H2,1-3H3
InChI key
JMJWCUOIOKBVNQ-UHFFFAOYSA-N
1 of 4
This Item | SCC461 | SCC291 | SCC109 |
---|---|---|---|
technique(s) cell culture | mammalian: suitable | technique(s) cell based assay: suitable, cell culture | mammalian: suitable, receptor binding assay: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
growth mode N/A | growth mode N/A | growth mode N/A | growth mode - |
storage temp. ≤ − 140°C | storage temp. −196°C | storage temp. ≤ − 140°C | storage temp. - |
shipped in dry ice | shipped in liquid nitrogen | shipped in dry ice | shipped in dry ice |
manufacturer/tradename Sigma-Aldrich | manufacturer/tradename Millipore | manufacturer/tradename Sigma-Aldrich | manufacturer/tradename - |
Application
- Sulfonyl hydroxamate derivatives for use as inhibitors of class ii fructosediphosphate aldolase
- Imidazole glycerol phosphate dehydratase inhibitors
- Competitive N-methyl-D-aspartate antagonists with analgesic activity
- Farnesyl phosphonate derivatives of phenylalanine for inhibition of farnesyl protein transferase
Reactant for condensation reactions with isobutyraldehyde
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service