About This Item
Recommended Products
assay
98%
mp
102-104 °C (lit.)
SMILES string
CC(=O)N1CCc2ccccc12
InChI
1S/C10H11NO/c1-8(12)11-7-6-9-4-2-3-5-10(9)11/h2-5H,6-7H2,1H3
InChI key
RNTCWULFNYNFGI-UHFFFAOYSA-N
General description
Application
- Reactant for preparation of triazolothiadiazepine dioxide derivatives
- Reactant for preparation of halo-substituted aromatic amides
- Reactant for preparation of [[(phenyl)piperazinyl]alkyl]indolyl]ethanone and [[[(phenoxyethyl)piperazinyl]alkyl]indolyl]ethanone derivatives as α1-adrenoreceptor antagonists
- Reactant for synthesis of naphthalenedione derivatives as antimycobacterial agents
- Reactant for regioselective ortho Suzuki-Miyaura coupling reaction
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service