Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

369403

Sigma-Aldrich

1,2-Propanediol dibenzoate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CO2CH2CH(CH3)O2CC6H5
CAS Number:
Molecular Weight:
284.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

refractive index

n20/D 1.545 (lit.)

bp

232 °C/12 mmHg (lit.)

mp

−3 °C (lit.)

density

1.16 g/mL at 25 °C (lit.)

SMILES string

CC(COC(=O)c1ccccc1)OC(=O)c2ccccc2

InChI

1S/C17H16O4/c1-13(21-17(19)15-10-6-3-7-11-15)12-20-16(18)14-8-4-2-5-9-14/h2-11,13H,12H2,1H3

InChI key

UMVMVEZHMZTUHD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

280.4 °F - closed cup

flash_point_c

138 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service