367257
(1S,2R)-(+)-trans-2-Phenyl-1-cyclohexanol
99%
Synonym(s):
(1S,2R)-trans-2-Phenyl-1-cyclohexanol
About This Item
Recommended Products
assay
99%
form
solid
optical activity
[α]20/D +58°, c = 7 in methanol
optical purity
ee: 98% (GLC)
bp
276-281 °C (lit.)
mp
64-66 °C (lit.)
SMILES string
O[C@H]1CCCC[C@@H]1c2ccccc2
InChI
1S/C12H16O/c13-12-9-5-4-8-11(12)10-6-2-1-3-7-10/h1-3,6-7,11-13H,4-5,8-9H2/t11-,12+/m1/s1
InChI key
AAIBYZBZXNWTPP-NEPJUHHUSA-N
Application
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service