Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

348333

Sigma-Aldrich

tert-Butyl diethylphosphonoacetate

95%

Synonym(s):

(Diethoxyphosphinyl)acetic acid 1,1-dimethylethyl ester, tert-Butyl (diethoxyphosphinyl)acetate, Diethyl tert-butoxycarbonylmethanephosphonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2P(O)CH2CO2C(CH3)3
CAS Number:
Molecular Weight:
252.24
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

liquid

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.431 (lit.)

bp

100-103 °C/1.5 mmHg (lit.)

density

1.074 g/mL at 25 °C (lit.)

functional group

ester
phosphonate

SMILES string

CCOP(=O)(CC(=O)OC(C)(C)C)OCC

InChI

1S/C10H21O5P/c1-6-13-16(12,14-7-2)8-9(11)15-10(3,4)5/h6-8H2,1-5H3

InChI key

NFEGNISFSSLEGU-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of ydroxymethylated dihydroxyvitamin D3 analogs via Wittig-Horner approach, as potential antitumor agents
  • Synthesis of phosphopeptide mimetic prodrugs targeted to Src homology 2 (SH2) domain of signal transducer and activator of transcription 3 (Stat 3)
  • One-pot synthesis of the DPP4-selective inhibitor ABT-341 by a four-component coupling mediated by a diphenylprolinol silyl ether
  • Bicyclic triaminophosphine-promoted stereoselective synthesis of a,ß-unsaturated esters, fluorides, and nitriles from aldehydes and ketones using Wadsworth-Emmons phosphonates
  • Preparation of amidopropenyl hydroxamates as human histone deacetylase (HDAC) inhibitors

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yosuke Kato et al.
Rapid communications in mass spectrometry : RCM, 34(7), e8636-e8636 (2019-10-30)
Ferulic acid (FA) is a standard matrix used for analyzing proteins. In this study, the ability of a halogenated FA to serve as an effective MALDI matrix was investigated. Various halogenated FAs were synthesized, and the characteristics and performance of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service