343617
Oxacyclododecan-2-one
98%
Synonym(s):
1-Oxa-2-cyclododecanone, 11-Undecanolide, Undecanoic ω-lactone
About This Item
Recommended Products
assay
98%
form
liquid
refractive index
n20/D 1.47 (lit.)
bp
124-126 °C/13 mmHg (lit.)
mp
2-3 °C (lit.)
density
0.992 g/mL at 25 °C (lit.)
functional group
ester
SMILES string
O=C1CCCCCCCCCCO1
InChI
1S/C11H20O2/c12-11-9-7-5-3-1-2-4-6-8-10-13-11/h1-10H2
InChI key
MVOSYKNQRRHGKX-UHFFFAOYSA-N
General description
Application
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
230.0 °F - closed cup
flash_point_c
110 °C - closed cup
ppe
Eyeshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service