Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

34310

Sigma-Aldrich

Methyl 2,3-dibromopropionate

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2BrCHBrCOOCH3
CAS Number:
Molecular Weight:
245.90
Beilstein/REAXYS Number:
1750190
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.514

bp

83-86 °C/10 mmHg (lit.)

density

1.944 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)C(Br)CBr

InChI

1S/C4H6Br2O2/c1-8-4(7)3(6)2-5/h3H,2H2,1H3

InChI key

ROXQOUUAPQUMLN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl 2,3-dibromopropionate is the methyl ester of 2,3-dibromopropanoic acid. It reacts with 2-acetamido-3-hydroxypyridine to give 2-substituted pyrido-oxazine.[1]

application

Methyl 2,3-dibromopropionate may be used in the preparation of methyl 2-azidoacrylate.[2] It may be used in the preparation of methyl (+)-(1′R, 2R) and (-)-(1′R, 2S)-1-(2-phenylethanol)aziridine-2-carboxylates.[3]

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regiospecific and Enantiospecific Ring Opening of Methyl (+)-(1'R, 2R)-and (-)-(1'R, 2S)-1-(2-phenylethanol) Aziridine-2-carboxylates.
Gnecco D, et al.
Molecules (Basel), 5(8), 998-1003 (2000)
Synthesis of substituted 3, 4-dihydro-2< i> H</i>-pyrido [3, 2-< i> b</i>][1, 4] oxazine as new scaffolds for potential bioactive compounds.
Henry N, et al.
Tetrahedron, 62(10), 2405-2412 (2006)
2H-Azirines as dipolarophiles.
Pinho e Melo TMVD, et al.
Tetrahedron Letters, 44(33), 6313-6315 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service