Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

340030

Sigma-Aldrich

2-Phenyl-1,3-dioxolane

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10O2
CAS Number:
Molecular Weight:
150.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

refractive index

n20/D 1.526 (lit.)

bp

80 °C/0.3 mmHg (lit.)

density

1.106 g/mL at 25 °C (lit.)

SMILES string

C1COC(O1)c2ccccc2

InChI

1S/C9H10O2/c1-2-4-8(5-3-1)9-10-6-7-11-9/h1-5,9H,6-7H2

InChI key

LYINTWKRUWVLBA-UHFFFAOYSA-N

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

208.4 °F - closed cup

flash_point_c

98.00 °C - closed cup

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service