Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

331015

Sigma-Aldrich

Pentafluoroiodoethane

97%

Synonym(s):

Iodopentafluoroethane, Perfluoroethyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CF2I
CAS Number:
Molecular Weight:
245.92
Beilstein/REAXYS Number:
1740331
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
Product 331015 is not currently sold in your country. Contact Technical Service

vapor pressure

738.9 mmHg ( 10 °C)

assay

97%

refractive index

n20/D 1.339 (lit.)

bp

12-13 °C (lit.)

density

2.085 g/mL at 25 °C (lit.)

functional group

fluoro
iodo

SMILES string

FC(F)(F)C(F)(F)I

InChI

1S/C2F5I/c3-1(4,5)2(6,7)8

InChI key

UXPOJVLZTPGWFX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a Sure/Pac cylinder and has a brass needle valve with a male 1/4" NPTF outlet thread installed. Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.

Compatible with the following:

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC

also commonly purchased with this product

Product No.
Description
Pricing

hose barb

Product No.
Description
Pricing

regulator

Product No.
Description
Pricing

pictograms

Gas cylinderExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Press. Gas Compr. Gas - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

supp_hazards

Storage Class

2A - Gases

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service