Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

30720

Sigma-Aldrich

2-Dodecanone

≥97.0% (GC)

Synonym(s):

Decyl methyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9COCH3
CAS Number:
Molecular Weight:
184.32
Beilstein/REAXYS Number:
1703135
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.433

bp

247 °C (lit.)

mp

17-20 °C

density

0.828 g/mL at 20 °C (lit.)

SMILES string

CCCCCCCCCCC(C)=O

InChI

1S/C12H24O/c1-3-4-5-6-7-8-9-10-11-12(2)13/h3-11H2,1-2H3

InChI key

LSKONYYRONEBKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Dodecanone was screened for its toxicity towards spider mites.

Application

2-Dodecanone was used in the synthesis of brushed block copolymers.

pictograms

Environment

hcodes

pcodes

Hazard Classifications

Aquatic Chronic 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

226.4 °F - closed cup

flash_point_c

108.00 °C - closed cup

ppe

Eyeshields, Gloves


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

George F Antonious et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 49(7), 527-531 (2014-05-13)
Glandular trichomes on the leaves of wild tomato, L. hirsutum f. hirsutum Mull, also known as Solanum habrochaites (Solanaceae), synthesize and accumulate high levels of methyl ketones (MKs). L. hirsutum accession LA 407, having high concentration of MKs, was grown
Hyun Jin Lee et al.
Pharmaceutical research, 30(8), 2077-2086 (2013-05-03)
To investigate the effects of small aliphatic pendent groups conjugated through an acid-sensitive linker to the core of brushed block copolymer micelles on particle properties. The brushed block copolymers were synthesized by conjugating five types of 2-alkanone (2-butanone, 2-hexanone, 2-octanone
Brian D Fitz et al.
Journal of chromatography. A, 1392, 82-90 (2015-03-31)
Low thermal mass gas chromatography (LTM-GC) was evaluated for rapid, high peak capacity separations with three injection methods: liquid, headspace solid phase micro-extraction (HS-SPME), and direct vapor. An Agilent LTM equipped with a short microbore capillary column was operated at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service