Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

303607

Sigma-Aldrich

3,3-Diethoxy-1-propyne

97%

Synonym(s):

Propargylaldehyde diethyl acetal, Propiolaldehyde diethyl acetal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2CHC≡CH
CAS Number:
Molecular Weight:
128.17
Beilstein/REAXYS Number:
1701566
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.412 (lit.)

bp

138-139.5 °C (lit.)

density

0.894 g/mL at 25 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

CCOC(OCC)C#C

InChI

1S/C7H12O2/c1-4-7(8-5-2)9-6-3/h1,7H,5-6H2,2-3H3

InChI key

RGUXEWWHSQGVRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3,3-Diethoxy-1-propyne was used in preparation of 3-boronoacrolein pinacolate, a heterodyne[1] and (E)-3-(tributylstannyl)-2-propenal.[2]

pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

87.8 °F - closed cup

flash_point_c

31 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Catalytic Asymmetric Addition of Polyfunctional Dialkylzincs to. beta.-Stannylated and. beta.-Silylated Unsaturated Aldehydes.
Ostwald R, et al.
The Journal of Organic Chemistry, 59(15), 4143-4153 (1994)
Xuri Gao et al.
Journal of the American Chemical Society, 125(31), 9308-9309 (2003-08-02)
This Communication reports the optimization of the first catalytic enantio- and diastereoselective hetero[4+2]/allylboration reaction to provide efficient access to alpha-hydroxyalkyl pyran derivatives. The key substrate 3-boronoacrolein pinacolate appears to be an exceptionally favorable heterodiene for use in Jacobsen's enantioselective reverse

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service