Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

301655

Sigma-Aldrich

4-Pyridylacetonitrile hydrochloride

98%

Synonym(s):

4-Pyridineacetonitrile hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N2 · HCl
CAS Number:
Molecular Weight:
154.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

267 °C (dec.) (lit.)

SMILES string

Cl[H].N#CCc1ccncc1

InChI

1S/C7H6N2.ClH/c8-4-1-7-2-5-9-6-3-7;/h2-3,5-6H,1H2;1H

InChI key

UBYQSEXKPQZPCN-UHFFFAOYSA-N

General description

Acylation reaction of 4-pyridylacetonitrile hydrochloride has been reported.[1]

Application

4-Pyridylacetonitrile hydrochloride was used in preparation of (2Z)-3-(4-hydroxyphenyl)-4-pyridin-4-ylacrylonitrile.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and X-ray structural analysis of (2Z)-3 (4-hydroxyphenyl)-2-pyridin-4-ylacrylonitrile and (2Z)-3-(3, 5-di-tert-butyl-4-hydroxyphenyl) 2-pyridin-4-ylacrylonitrile.
Helvenston MC, et al.
Journal of Chemical Crystallography, 35(2), 113-118 (2005)
Acylation and other reactions of 2-and 4-pyridylacetonitrile.
Gutsche CD and Voges HW.
The Journal of Organic Chemistry, 32(9), 2685-2689 (1967)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service