Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300683

Sigma-Aldrich

α-1,2,3,4,5,6-Hexachlorocyclohexane

99%

Synonym(s):

α-BHC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H6Cl6
CAS Number:
Molecular Weight:
290.83
Beilstein/REAXYS Number:
7073451
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

99%

form

solid

mp

156-161 °C (lit.)

SMILES string

Cl[C@@H]1[C@H](Cl)[C@@H](Cl)[C@H](Cl)[C@@H](Cl)[C@H]1Cl

InChI

1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H/t1-,2-,3-,4-,5+,6+/m1/s1

InChI key

JLYXXMFPNIAWKQ-SHFUYGGZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-1,2,3,4,5,6-Hexachlorocyclohexane is a racemic compound with anticonvulsant and tremorogenic activities and its accumulation in cerebral white over grey matter in rats was confirmed. Administration of α -1,2,3,4,5,6-hexachlorocyclohexane (α-HCH) to rats decreased the utilization of [2-14C]orotic acid for the synthesis of liver cytidine nucleotides.

signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Seifert et al.
Toxicology, 7(2), 155-161 (1977-04-01)
Administration of alpha-1,2,3,4,5,6-hexachlorocyclohexane (alpha-HCH) to rats decreased the utilization of [2-14C]orotic acid for the synthesis of liver cytidine nucleotides. The specific radioactivities of uridine components of the acid-soluble pool and rRNA increased during the first hours of treatment with the
J Portig et al.
Xenobiotica; the fate of foreign compounds in biological systems, 19(1), 123-130 (1989-01-01)
1. Whole-body autoradiographic evidence that alpha-1,2,3,4,5,6-hexachlorocyclohexane (alpha-HCH), a racemic compound with anticonvulsant and tremorogenic activities, accumulates in cerebral white over grey matter was confirmed in rats using unlabelled compound and g.l.c. determination in tissue samples. 2. Accumulation in, and retention
Pernilla Carlsson et al.
Journal of environmental monitoring : JEM, 14(5), 1437-1443 (2012-04-07)
A pilot study is presented evaluating selected chlorinated pesticides as chemical tracers for water masses in a sub-Arctic fjord system (Godthåbsfjord, western Greenland). Polyoxymethylene (POM) based passive water samplers were deployed during summer-autumn 2010. The levels of the analysed chlorinated
Sung-Deuk Choi et al.
Environmental science & technology, 45(20), 8834-8841 (2011-09-13)
An understanding of the factors that control the time trends of persistent organic pollutants (POPs) in the environment is required to evaluate the effectiveness of emission reductions and to predict future exposure. Using a regional contaminant fate model, CoZMo-POP 2
M Pućko et al.
Environmental science & technology, 46(21), 11862-11869 (2012-10-09)
During the summer of 2009, we sampled 14 partially refrozen melt ponds and the top 1 m of old ice in the pond vicinity for α-hexachlorocyclohexane (α-HCH) concentrations and enantiomer fractions (EFs) in the Beaufort Sea. α-HCH concentrations were 3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service