Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

300284

Sigma-Aldrich

Di(ethylene glycol) hexyl ether

97%

Synonym(s):

2-(2-Hexyloxyethoxy)ethanol, C6E2, Hexyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
190.28
Beilstein/REAXYS Number:
1743959
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.4381 (lit.)

bp

260 °C (lit.)

mp

−40 °C (lit.)

density

0.935 g/mL at 25 °C (lit.)

SMILES string

CCCCCCOCCOCCO

InChI

1S/C10H22O3/c1-2-3-4-5-7-12-9-10-13-8-6-11/h11H,2-10H2,1H3

InChI key

GZMAAYIALGURDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Dermal - Eye Dam. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

253.4 °F

flash_point_c

123 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

B Ballantyne et al.
Journal of applied toxicology : JAT, 21(6), 449-460 (2001-12-18)
Diethylene glycol monohexyl ether (DEGHE; CAS no. 112-59-4), an industrial chemical, was investigated for the potential to produce genotoxic effects using three in vitro and two in vivo tests. No mutagenic activity occurred in either the absence or presence of
B Ballantyne et al.
Veterinary and human toxicology, 29(5), 361-366 (1987-10-01)
Ethylene glycol monohexyl ether (EGHE) and diethylene glycol monohexyl ether (DGHE) are glycol ethers used as industrial solvents and coating materials, and whose acute handling hazards were investigated. Acute peroral LD50 values in the rat were for EGHE 1.67 ml/kg

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service