About This Item
Recommended Products
assay
96%
refractive index
n20/D 1.543 (lit.)
bp
228-230 °C (lit.)
density
1.15 g/mL at 25 °C (lit.)
functional group
ketone
SMILES string
CC(=O)c1sc(C)nc1C
InChI
1S/C7H9NOS/c1-4-7(5(2)9)10-6(3)8-4/h1-3H3
InChI key
BLQOKWQUTLNKON-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
10 - Combustible liquids
wgk_germany
WGK 3
flash_point_f
219.2 °F - closed cup
flash_point_c
104 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service