About This Item
Recommended Products
vapor density
0.14 (vs air)
Quality Level
assay
≥99.995%
impurities
≤5 ppm oxygen
≤5 ppm water
bp
-268.934 °C (lit.)
−268.9 °C (lit.)
mp
-272.2 °C (lit.)
SMILES string
[He]
InChI
1S/He
InChI key
SWQJXJOGLNCZEY-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Compatible with the following:
- Aldrich® lecture-bottle station systems
- Aldrich® lecture-bottle gas regulators
Other Notes
Legal Information
also commonly purchased with this product
control valve
hose barb
purge valve
recommended
signalword
Warning
hcodes
pcodes
Hazard Classifications
Press. Gas Compr. Gas
Storage Class
2A - Gases
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service