29438
(R)-2-Oxiranylanisole
≥97.0% (sum of enantiomers, GC)
Synonym(s):
(R)-α-(2-Oxiranyl)anisole, (R)-Glycidyl phenyl ether, (R)-Phenoxymethyloxirane
About This Item
Recommended Products
assay
≥97.0% (sum of enantiomers, GC)
SMILES string
C1O[C@H]1COc2ccccc2
InChI
1S/C9H10O2/c1-2-4-8(5-3-1)10-6-9-7-11-9/h1-5,9H,6-7H2/t9-/m0/s1
Inchi Key
FQYUMYWMJTYZTK-VIFPVBQESA-N
Application
- Neuroprotective small molecules
- Piperidinol analogs for studies of anti-tuberculosis activity
- 2-Substituted 3,4-dihydro-2H-1,4-benzoxazines via cyclization of hydroxyl sulfonamides
- Bicyclic azetidines via ring opening, esterification, chlorination and intramolecular alkylation
- Bromoalkanes via ring opening and addition across C-O bonds
Involved in biological studies as a substrate for Bacillus-produced enantioselective eposide hydrolase
signalword
Danger
Hazard Classifications
Acute Tox. 4 Dermal - Carc. 1B - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 3
flash_point_f
159.8 °F - closed cup
flash_point_c
71 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service